3-(Trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Trifluoromethoxyaniline

A165799-100G A165799-10G A165799-1G A165799-25G A165799-5G
103844-252EA 6 USD
103844-252 77138-308 77138-310 77138-312 77300-800 77320-472
3-(Trifluoromethoxy)aniline 98%
3-(Trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 195.9 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00041511
CAS Number: 1535-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR