3-Thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one 95%

Supplier: Ambeed
Warning

A141669-100MG A141669-250MG A141669-1G A141669-5G A141669-25G
103803-036EA 47 USD
103803-036 76767-790 76767-792 77299-238 77299-240 77624-174
3-Thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one 95%
3-Thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

Precaución: for Research Use Only

Formula: C₃H₃N₃OS
Boiling Pt: 373.1 °C (760 mm Hg)
Flash Pt: 179.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00047460
CAS Number: 626-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR