3-Thienylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: Thiophene-3-acetic acid, 3-Thiopheneacetic acid

A379925-1G A379925-25G A379925-5G A379925-100G A379925-10G A379925-500G
103826-654EA 9 USD
103826-654 103837-690 76841-006 76841-008 76841-010 77115-490 77115-492 77115-494
3-Thienylacetic acid 98%
3-Thienylacetic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 274.3 °C (760 mm Hg)
Melting Pt: 77…81 °C
Flash Pt: 119.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS Number: 6964-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR