3-Sulfamoylbenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A126577-250MG
A126577-1G
A126577-5G
103800-838EA
10
USD
103800-838
77108-888
77313-064
77313-066
3-Sulfamoylbenzoic acid 95%
3-Sulphamoylbenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₄S Boiling Pt: 476.1 °C (760 mm Hg) Melting Pt: 251…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD01122318 CAS Number: 636-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...