3-Pyrrolidinone hydrochloride 97%

Supplier: Ambeed
Warning

A207305-1G A207305-25G A207305-5G A207305-10G
103817-782EA 17 USD
103817-782 103848-462 77086-730 77086-732 77086-734 77287-826
3-Pyrrolidinone hydrochloride 97%
3-Pyrrolidinone hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO
MW: 121.5654 g/mol
Boiling Pt: 197.8 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 73.4 °C
Storage Temperature: Freezer
MDL Number: MFCD02183811
CAS Number: 3760-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR