3-Pyridylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol

A345889-25G A345889-100G A345889-10G A345889-500G A345889-5G
103836-918EA 5 USD
103836-918 76839-988 77290-592 77290-594 77290-596 77306-570
3-Pyridylmethanol 98%
3-Pyridylmethanol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 266 °C (760 mm Hg)
Melting Pt: 7.7 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006407
CAS Number: 100-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR