Nicotinaldehyde 97%

Supplier: Ambeed
Danger

Synonyms: Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde

A698729-5G A698729-10G A698729-25G
103840-620EA 5 USD
103840-620 76844-850 77104-914 77308-144
Nicotinaldehyde 97%
Nicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 209.9 °C (760 mm Hg)
Melting Pt: 8 °C
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006382
CAS Number: 500-22-1
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR