3-Propoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A281415-10G A281415-250MG A281415-1G A281415-5G
103813-548EA 17 USD
103813-548 103853-498 77250-506 77250-508 77305-390 77305-392
3-Propoxyphenylboronic acid 98%
3-Propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.0100 g/mol
Boiling Pt: 339.4 °C (760 mm Hg)
Melting Pt: 99…106 °C
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD03427190
CAS Number: 149557-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR