3-Piperazin-1-yl-pyridazine 97%

Supplier: Ambeed
Warning

A696123-100MG A696123-250MG
103840-606EA 704 USD
103840-606 77198-784 77198-786
3-Piperazin-1-yl-pyridazine 97%
3-Piperazin-1-yl-pyridazine

Precaución: for Research Use Only

Formula: C8H12N4
MW: 164.2100 g/mol
Boiling Pt: 381.1±22.0 °C (760 mm Hg)
Flash Pt: 184.268 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01563781
CAS Number: 51047-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR