3-Phenylpyrrolidine 98%

Supplier: Ambeed
Warning

A281057-5G A281057-100MG A281057-250MG A281057-1G A281057-25G
103813-534EA 36 USD
103813-534 103853-486 76837-182 77114-066 77114-068 77305-374 77305-376
3-Phenylpyrrolidine 98%
3-Phenylpyrrolidine

Precaución: for Research Use Only

Formula: C10H13N
Boiling Pt: 243.4 °C (760 mm Hg)
Flash Pt: 103.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01838169
CAS Number: 936-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR