2-Hydroxy-3-phenylpyridine 98%

Supplier: Ambeed
Warning

A121442-100MG A121442-250MG
103800-076EA 235 USD
103800-076 77258-118 77258-120
2-Hydroxy-3-phenylpyridine 98%
2-Hydroxy-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₉NO
Boiling Pt: 417.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114134
CAS Number: 24228-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR