3-Phenylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A106563-100MG A106563-250MG A106563-1G
103797-042EA 550 USD
103797-042 103822-796 77148-506 77148-508 77309-654
3-Phenylpyridin-2-amine 95%
3-Phenylpyridin-2-amine

Precaución: for Research Use Only

Formula: C11H10N2
Boiling Pt: 320.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114119
CAS Number: 87109-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR