3-Phenylpropiolaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Phenylpropargyl aldehyde

A192621-1G A192621-25G A192621-5G A192621-10G A192621-100G
103816-466EA 5 USD
103816-466 103847-076 77084-946 77084-948 77084-950 77164-328 77683-512
3-Phenylpropiolaldehyde 98%
3-Phenylpropiolaldehyde

Precaución: For research use only.

Formula: C₉H₆O
MW: 130.15 g/mol
Boiling Pt: 246.4 °C (760 mm Hg)
Flash Pt: 202 °C
Storage Temperature: Freezer
MDL Number: MFCD00006995
CAS Number: 2579-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR