3-Phenyl-2-propyn-1-ol 98%

Supplier: Ambeed
Warning

A276643-5G A276643-100G A276643-10G A276643-1G A276643-25G
103813-314EA 9 USD
103813-314 103853-250 77093-116 77289-560 77289-562 77305-242 77305-244
3-Phenyl-2-propyn-1-ol 98%
3-Phenyl-2-propyn-1-ol

Precaución: for Research Use Only

Formula: C₉H₈O
Boiling Pt: 241.8 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR