3-Phenyloxetan-3-ol 98%

Supplier: Ambeed
Warning

A195817-100MG A195817-250MG A195817-1G
103816-788EA 251 USD
103816-788 77012-496 77012-498 77085-346
3-Phenyloxetan-3-ol 98%
3-Phenyloxetan-3-ol

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.175 g/mol
Boiling Pt: 296.9 °C (760 mm Hg)
Melting Pt: 55…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18909281
CAS Number: 699-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR