3-Phenylbenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 3-(Bromomethyl)biphenyl, m-(Bromomethyl)biphenyl

A427979-10G A427979-1G A427979-25G A427979-5G
103827-494EA 25 USD
103827-494 103838-548 76725-300 76842-214 76842-216 76842-218
3-Phenylbenzyl bromide 97%
3-Phenylbenzyl bromide

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Freezer
MDL Number: MFCD03452790
CAS Number: 14704-31-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR