3-Hydroxy-3-phenylazetidine hydrochloride 97%

Supplier: Ambeed
Warning

A115352-100MG A115352-250MG A115352-1G A115352-5G
103798-554EA 89 USD
103798-554 103824-408 76982-936 76982-938 77297-366 77297-368
3-Hydroxy-3-phenylazetidine hydrochloride 97%
3-Hydroxy-3-phenylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD22570262
CAS Number: 550370-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR