3-Phenyl-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Phenyl-1-Propanol

A424112-1000G A424112-100G A424112-500G A424112-25G
103810-320EA 85 USD
103810-320 76842-132 77291-444 77291-446 77307-222
3-Phenyl-1-propanol 98%
3-Phenyl-1-propanol

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CH₂CH₂OH
Boiling Pt: 119…121 °C (760 mm Hg)
Melting Pt: 18 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR