3-Phenyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

A104401-100G A104401-10G A104401-1G A104401-5G A104401-25G
103796-704EA 5 USD
103796-704 103822-418 77283-204 77283-206 77309-286 77309-288 77592-108
3-Phenyl-1H-pyrazole 98%
3-Phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18
Boiling Pt: 339.1 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 163.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00159654
CAS Number: 2458-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR