3-Oxo-2,3-dihydro-1H-indene-5-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-indan-5-carbonitrile

A460940-1G A460940-5G A460940-250MG
103827-920EA 24 USD
103827-920 103838-998 76842-878 76842-880 77116-288
3-Oxo-2,3-dihydro-1H-indene-5-carbonitrile 97%
3-Oxo-2,3-dihydro-1H-indene-5-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.169 g/mol
Boiling Pt: 312.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02179291
CAS Number: 69975-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR