3-Oxocyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A112722-100G A112722-10G A112722-25G A112722-500G A112722-5G A112722-1G
103798-086EA 5 USD
103798-086 77094-074 77094-076 77094-078 77094-080 77094-082 77310-742
3-Oxocyclobutanecarboxylic acid 98%
3-Oxocyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C5H6O3
MW: 114.10 g/mol
Boiling Pt: 296.5 °C (760 mm Hg)
Melting Pt: 58…60 °C
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00100900
CAS Number: 23761-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR