Benzoylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Phenyl-3-oxopropanenitrile, 3-Oxo-3-phenylpropanenitrile

A273670-100G A273670-10G A273670-500G A273670-25G A273670-5G
103853-076EA 6 USD
103853-076 76692-888 76692-890 76692-892 77092-880 77092-882
Benzoylacetonitrile 98%
Benzoylacetonitrile

Precaución: for Research Use Only

Formula: (C₆H₅)C(O)CH₂CN
MW: 145.16 g/mol
Boiling Pt: 160 °C (760 mm Hg)
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001942
CAS Number: 614-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR