3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic acid

A409981-1G A409981-100MG A409981-250MG
103827-206EA 46 USD
103827-206 76841-766 77291-272 77291-274
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid 95%
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03036647
CAS Number: 134997-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR