3-Oxo-3-(2-pyridinyl)propanenitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3-(pyridin-2-yl)propanenitrile

A109017-250MG A109017-25G
103797-426EA 81 USD
103797-426 103823-214 77270-740 77270-742
3-Oxo-3-(2-pyridinyl)propanenitrile 97%
3-Oxo-3-(2-pyridinyl)propanenitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Boiling Pt: 312.705 °C (760 mm Hg)
Melting Pt: 97…99 °C
Flash Pt: 142.919 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656806
CAS Number: 54123-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR