3-Oxetanamine 97%

Supplier: Ambeed
Danger

Synonyms: 3-amino-oxetane, 3-Aminooxetane

A243650-1G A243650-10G
103811-532EA 6 USD
103811-532 103851-354 77090-558 77568-364
3-Oxetanamine 97%
3-Oxetanamine

Precaución: For research use only.

Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 98.8 °C (760 mm Hg)
Flash Pt: 54.4 °C
Storage Temperature: Freezer
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1992
ADR: 3,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR