3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A246010-100MG A246010-250MG A246010-1G A246010-5G
103811-680EA 159 USD
103811-680 103851-510 76976-584 76976-586 77090-784 77090-786
3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid 98%
3-Oxabicyclo[3.1.0]hexane-6-carboxylic acid

Precaución: for Research Use Only

Formula: C6H8O3
MW: 128.13 g/mol
Boiling Pt: 281.2 °C (760 mm Hg)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD13193581
CAS Number: 693248-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR