3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride 97%

Supplier: Ambeed
Warning

A155297-50MG A155297-250MG
103804-740EA 365 USD
103804-740 76812-418 77611-476
3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride 97%
3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉NO·ClH
MW: 135.592 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193583
CAS Number: 693248-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR