3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride 97%

Supplier: Ambeed
Warning

A258430-250MG A258430-1G A258430-25G A258430-5G
103812-374EA 65 USD
103812-374 103852-248 76976-864 77304-734 77304-736 77304-738
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride 97%
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride

Precaución: for Research Use Only

Formula: C6H12ClNO
Storage Temperature: Ambient
MDL Number: MFCD18432562
CAS Number: 904316-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR