3'-O-tert-Butyldimethylsilyl-5'-O-DMT-N2-isobutyrylguanosine 2'-CE phosphoramidite 95%

Supplier: Ambeed
Warning

A159723-100MG A159723-250MG A159723-1G
103805-278EA 180 USD
103805-278 103843-504 77191-038 77191-040 77319-406
3'-O-tert-Butyldimethylsilyl-5'-O-DMT-N2-isobutyrylguanosine 2'-CE phosphoramidite 95%
3'-O-tert-Butyldimethylsilyl-5'-O-DMT-N2-isobutyrylguanosine 2'-CE phosphoramidite

Precaución: Sealed in dry, at 2…8°C

Formula: C₅₀H₆₈N₇O₉PSi
MW: 970.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30475539
CAS Number: 1445905-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR