3-Nitrophthalimide 98%

Supplier: Ambeed
Warning

A134236-100G A134236-500G A134236-1000G A134236-25G
103808-482EA 95 USD
103808-482 77190-130 77190-132 77314-720 77314-722
3-Nitrophthalimide 98%
3-Nitrophthalimide

Precaución: for Research Use Only

Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00041852
CAS Number: 603-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR