3-Nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

A184709-25G A184709-5G A184709-100G A184709-10G A184709-1G
103846-338EA 10 USD
103846-338 77083-956 77083-958 77111-542 77111-544 77111-546
3-Nitrophenylboronic acid 98%
3-Nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BNO₄
Melting Pt: 284…285 °C
Flash Pt: 173.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007193
CAS Number: 13331-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR