3-Nitrobenzylamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Nitrobenzylammonium hydrochloride
A239387-100G
A239387-10G
A239387-25G
A239387-5G
103851-084EA
51
USD
103851-084
77193-286
77193-288
77304-156
77304-158
3-Nitrobenzylamine hydrochloride 97%
3-Nitrobenzylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₉ClN₂O₂ MW: 188.62 g/mol Boiling Pt: 294 °C (760 mm Hg) Melting Pt: 229…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012858 CAS Number: 26177-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...