3-Nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A504175-25G A504175-10G A504175-1G A504175-5G
103839-416EA 6 USD
103839-416 76843-390 77004-710 77004-712 77307-628
3-Nitrobenzenesulfonamide 98%
3-Nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007935
CAS Number: 121-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR