3-Nitro-o-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine

A156894-250MG A156894-25G A156894-1G A156894-5G
103804-934EA 24 USD
103804-934 103843-136 77110-528 77110-530 77318-942 77318-944
3-Nitro-o-phenylenediamine 97%
3-Nitro-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Boiling Pt: 369.7 °C (760 mm Hg)
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00007722
CAS Number: 3694-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR