3'-Nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: m-Nitroacetophenone

A729569-1000G A729569-100G A729569-500G A729569-25G
103810-542EA 52 USD
103810-542 76845-006 77118-096 77118-098 77118-100 77308-222
3'-Nitroacetophenone 98%
3'-Nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 202
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR