5-Nitro-α,α,α-trifluoro-m-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-5-(trifluoromethyl)benzoic acid

A314722-1G A314722-100G A314722-10G A314722-25G A314722-500G A314722-5G
103825-044EA 5 USD
103825-044 103835-968 76838-788 77290-198 77290-200 77290-202 77290-204 77306-118
5-Nitro-α,α,α-trifluoro-m-toluic acid 97%
5-Nitro-α,α,α-trifluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
Boiling Pt: 309.8 °C (760 mm Hg)
Melting Pt: 128 °C
Flash Pt: 65.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00024509
CAS Number: 328-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR