3-Nitro-5-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A251815-5G A251815-250MG A251815-1G A251815-10G
103812-010EA 8 USD
103812-010 103851-850 77091-246 77098-690 77304-564 77685-715
3-Nitro-5-(trifluoromethyl)aniline 97%
3-Nitro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MW: 206.12
Boiling Pt: 280.7 °C (760 mm Hg)
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00061229
CAS Number: 401-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR