3-Nitropyrrole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-1H-pyrrole

A170763-100MG A170763-250MG A170763-25G A170763-1G A170763-5G
103806-540EA 44 USD
103806-540 103844-816 77286-842 77286-844 77286-846 77301-074 77301-076
3-Nitropyrrole 98%
3-Nitropyrrole

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 267.9 °C (760 mm Hg)
Melting Pt: 100 °C
Flash Pt: 115.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00059714
CAS Number: 5930-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR