3-Methylquinoxaline-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-2-quinoxalinecarboxylic acid

A125707-250MG A125707-1G
103800-712EA 224 USD
103800-712 76996-086 77312-862
3-Methylquinoxaline-2-carboxylic acid 97%
3-Methylquinoxaline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Boiling Pt: 347.1 °C (760 mm Hg)
Melting Pt: 168…170 °C
Flash Pt: 163.7 °C
Storage Temperature: Freezer
MDL Number: MFCD03840415
CAS Number: 74003-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR