3-Hydroxy-3-methylpyrrolidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methylpyrrolidin-3-ol hydrochloride
A596807-100MG
A596807-10G
A596807-250MG
A596807-1G
A596807-5G
A596807-25G
103828-954EA
67
USD
103828-954
103840-092
76830-878
76830-880
76830-882
76844-242
77307-908
77886-445
3-Hydroxy-3-methylpyrrolidine hydrochloride 97%
3-Hydroxy-3-methylpyrrolidine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD13191768 CAS Number: 921592-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...