3-Methyl-1H-pyrazole 98%

Supplier: Ambeed
Danger

A101522-100G A101522-10G A101522-25G A101522-500G A101522-5G
103821-694EA 5 USD
103821-694 76994-512 76994-514 76994-516 76994-518 76994-520 77297-142
3-Methyl-1H-pyrazole 98%
3-Methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₆N₂
Boiling Pt: 204 °C (760 mm Hg)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005240
CAS Number: 1453-58-3
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR