2-Formyl-3-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde

A446074-1G A446074-100G A446074-10G
103827-736EA 11 USD
103827-736 103838-800 76842-594 76979-432 76979-434
2-Formyl-3-picoline 97%
2-Formyl-3-picoline

Precaución: For research use only

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 207 °C (760 mm Hg)
Flash Pt: 73.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181144
CAS Number: 55589-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR