3-Methyloxindole 97%

Supplier: Ambeed
Warning

A389351-100G A389351-10G A389351-250MG A389351-1G A389351-25G A389351-5G
103826-846EA 15 USD
103826-846 103837-886 77278-600 77278-602 77278-604 77306-988 77306-990 77306-992
3-Methyloxindole 97%
3-Methyloxindole

Precaución: for Research Use Only

Formula: C₉H₉NO
Boiling Pt: 279.3 °C (760 mm Hg)
Melting Pt: 117…121 °C at N/A mmHg
Flash Pt: 157.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04037370
CAS Number: 1504-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR