3-Methyloxetane-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-3-oxetanecarboxylic acid

A335087-1G A335087-25G A335087-5G A335087-10G A335087-250MG
103825-682EA 17 USD
103825-682 103836-652 76839-658 76839-660 76839-662 76978-320 76978-322
3-Methyloxetane-3-carboxylic acid 98%
3-Methyloxetane-3-carboxylic acid

Precaución: For research use only

Formula: C₅H₈O₃
MW: 116.116 g/mol
Boiling Pt: 223.5 °C (760 mm Hg)
Melting Pt: 58…60 °C
Flash Pt: 98.5 °C
Storage Temperature: Freezer
MDL Number: MFCD09842435
CAS Number: 28562-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR