3-Methyloxetan-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-3-methyloxetane

A147112-250MG A147112-1G
103803-740EA 54 USD
103803-740 103841-836 76812-010 77317-196
3-Methyloxetan-3-ol 97%
3-Methyloxetan-3-ol

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 88.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18909291
CAS Number: 162816-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR