3-Amino-3-methyloxetane 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyloxetan-3-amine, 3-Methyl-3-oxetanamine

A148892-100MG A148892-250MG A148892-1G A148892-25G
103803-976EA 42 USD
103803-976 103842-096 76973-012 76973-014 77299-692 77317-564
3-Amino-3-methyloxetane 98%
3-Amino-3-methyloxetane

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 101.8 °C (760 mm Hg)
Flash Pt: 96.6 °C
Storage Temperature: Freezer
MDL Number: MFCD11111713
CAS Number: 874473-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR