3-Methylmorpholine 97%
Supplier: Ambeed
|
Danger
|
A116362-100MG
A116362-250MG
A116362-5G
A116362-1G
103798-720EA
14
USD
103798-720
103824-580
77271-216
77271-218
77297-454
77311-398
3-Methylmorpholine 97%
3-Methylmorpholine
Precaución: for Research Use Only
Formula:
C5H11NO MW: 101.1469 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09054773 CAS Number: 42185-06-8 UN: 2734 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...