3-Methyl-5-isoxazolecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylisoxazole-5-carboxylic acid

A391806-1G A391806-100G A391806-10G A391806-250MG
103826-888EA 51 USD
103826-888 103837-926 76841-322 77278-616 77278-618 77278-620
3-Methyl-5-isoxazolecarboxylic acid 95%
3-Methyl-5-isoxazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.10 g/mol
Flash Pt: 142.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00464222
CAS Number: 4857-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR