3-Methyl-4-isoxazolecarboxylic acid 98%

Supplier: Ambeed
Warning

A157518-10G A157518-250MG A157518-1G A157518-5G A157518-100MG
103805-012EA 30 USD
103805-012 103843-212 77011-052 77011-054 77319-040 77319-042 77890-030
3-Methyl-4-isoxazolecarboxylic acid 98%
3-Methyl-4-isoxazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
Boiling Pt: 303.6 °C (760 mm Hg)
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD01318161
CAS Number: 17153-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR