3-Methylisonicotinic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methyl-4-pyridinecarboxylic acid
A110383-10G
A110383-250MG
A110383-25G
A110383-1G
A110383-5G
103797-658EA
21
USD
103797-658
103823-464
77270-846
77270-848
77270-850
77310-260
77310-262
3-Methylisonicotinic acid 97%
3-Methylisonicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ MW: 137.1360 g/mol Melting Pt: 240…244 °C Flash Pt: 188.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128870 CAS Number: 4021-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...